主要文件
GBL805025
Grace Bio-Labs PATH® protein microarray slides, no barcode
nitrocellulose (on glass slide), external size 25 mm × 75 mm, pack of 20 ea
别名:
non-porous nitrocellulose film slide
登录查看公司和协议定价
选择尺寸
20 EA
$1,360.00
$1,360.00
请联系客服了解存货情况
选择尺寸
变更视图
20 EA
$1,360.00
About This Item
UNSPSC代码:
41100000
NACRES:
NB.15
$1,360.00
请联系客服了解存货情况
物料
nitrocellulose (on glass slide)
包装
pack of 20 ea
制造商/商品名称
Grace Bio-Labs 805025
外部尺寸
25 mm × 75 mm
一般描述
The Grace Bio-Labs PATH protein microarray slides are a non-porous nitrocellulose film slides.
Benefits:
  • Low and consistant fluorescent backgrounds
  • Higher protein binding
  • Superior signal-to-noise
法律信息
PATH is a registered trademark of Courtagen Life Sciences, Inc.
历史批次信息供参考:
分析证书(COA)