Merck
EP5804706005
Eppendorf® 4 Falcon Buckets
each for 2 x 50 mL Falcon tubes for swing bucket rotor A-4-44
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103910
NACRES:
NB.82
特點
each for 2 x 50 mL Falcon tubes for swing bucket rotor A-4-44
包裝
set of 4 pieces
製造商/商標名
5804706005
Eppendorf® 5804706005
Eppendorf®
技術
centrifugation: suitable
法律資訊
Eppendorf is a registered trademark of Eppendorf AG