EP4922608003
1 of 10
Supelco
20619
分子筛 5A 除湿器
BR705880
BRAND® Transferpette® S 移液器
20298
Molecular Sieve 5A Water Vapor Trap Refill
Z564222
FRIO-Temp® digital thermometer
Avanti
840537L
18:1 SQDG(d18)
23909
OMI® 纯化器管
20618
Sigma-Aldrich
Z325155
BRAND® BLAUBRAND® ETERNA volumetric pipette, calibrated To Deliver
Z740537
KDS Legato™ 200 series syringe pump, infuse only
23991
includes piston, cylinder, tip spring, o ring
100 μL
yellow