主要文件
EP022639903
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-6-30, aerosol-tight, to hold, 6 x 15/50 mL conical tubes, lid, including software upgrade (required for some older models)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103909
如需咨询该产品EP022639903,请联系默克当地办事处或经销商。 联系技术服务部门
物料
aluminum
特点
RCF 20,133 × g
to hold 6 x 15/50 mL conical tubes, lid, including software upgrade (required for some older models)
aerosol-tight
包装
pack of 1 ea
制造商/商品名称
Eppendorf® 022639903
相容性
5804/5804R & 5810/5810R
FA-45-6-30
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Rotor FA-45-6-30, for 6 × 15/50 mL conical tubes, including software upgrade (required for some older models)
  • High-speed rotor for applications in tubes from 5 – 50 mL1)
  • Max. speed 20,133 × g (12,100 rpm) in Centrifuge 5810 R or 16,639 × g (11,000 rpm) in Centrifuge 5804/5804 R/58102)
  • Eppendorf QuickLock® rotor lid for fast lid locking
  • Aerosol-tight3) for safe centrifugation of hazardous samples
  • Low rotor weight (3.2 kg) for easy lifting
  • Anodized aluminum for high chemical resistance
  • 45° borehole angle minimizes pellets smear
  • Rotor, lid and adapters are autoclavable (121 °C, 20 min)
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information.
2) Rotors F-34-6-38 and FA-45-6-30 maintain 4 °C at 11,000 rpm
3) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain
特点和优势
  • High-speed rotor for applications in tubes from 5 – 50 mL1)
  • Max. speed 20,133 × g (12,100 rpm) in Centrifuge 5810 R or 16,639 × g (11,000 rpm) in Centrifuge 5804/5804 R/58102)
  • Eppendorf QuickLock® rotor lid for fast lid locking
  • Aerosol-tight3) for safe centrifugation of hazardous samples
  • Low rotor weight (3.2 kg) for easy lifting
  • Anodized aluminum for high chemical resistance
  • 45° borehole angle minimizes pellets smear
  • Rotor, lid and adapters are autoclavable (121 °C, 20 min)
  • 1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information.
  • 2) Rotors F-34-6-38 and FA-45-6-30 maintain 4 °C at 11,000 rpm
  • 3) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain
法律信息
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)