主要文件
CLS999938
Corning® phenolic cap with white rubber liner
38-400 thread
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
物料
black phenolic cap
rubber liner
包装
case of 144 (4 x 36 pack)
高度
12.5 mm
配件
38-400 thread
正在寻找类似产品? 访问 产品对比指南
一般描述
These reusable Corning caps are fabricated specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving. Caps have GPI 38-400 threads and are provided with a glued-in white rubber liner.
法律信息
Corning is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)