主要文件
CLS402648
Corning® Modified Impeller Assembly (for use with probes) for Magnetically-Driven Proculture® Spinner Flask
fits flask, 3 L
别名:
equipment replacement accessory
登录查看公司和协议定价
选择尺寸
1 EA
$337.00
$337.00
预计发货时间2025年5月15日详情
选择尺寸
变更视图
1 EA
$337.00
About This Item
UNSPSC代码:
41121804
NACRES:
NB.43
$337.00
预计发货时间2025年5月15日详情
物料
stainless steel
无菌性
non-sterile
特点
fits flask 3 L
制造商/商品名称
Corning 402648
正在寻找类似产品? 访问 产品对比指南
一般描述
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 3L Proculture Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
法律信息
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
历史批次信息供参考:
分析证书(COA)