主要文件
CLS3121250CO
Pyrex® large top desiccators with 24/29 standard taper stopcock
replacement cover for 10.5L large top desiccator w/ 24/29 standard taper stopcock, 1/cs
别名:
glass desiccator cover, glass desiccator lid
登录查看公司和协议定价
选择尺寸
1 EA
$429.00
$429.00
国内现货,预计发货时间2025年4月28日详情
选择尺寸
变更视图
1 EA
$429.00
About This Item
UNSPSC代码:
41104510
NACRES:
NB.43
$429.00
国内现货,预计发货时间2025年4月28日详情
物料
borosilicate glass (borosilicate glass code 7740)
特点
reusable
包装
case of 1
pack of 1
制造商/商品名称
Corning 3121-250CO
高度
109 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
These PYREX desiccator covers are replacements for 10.5L desiccators CLS3121250. They require a replacement 24/29 Standard Taper stopcock with a tooled hose connection. The plug material is PTFE.
法律信息
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)