主要文件
BMSW100510
Sigma-Aldrich
Accuris M1 Grade Calibration Weight
size 10 g, stainless steel
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NB.71
物料
stainless steel
包装
pack of 1 ea
制造商/商品名称
Benchmark Scientific W1005-10
尺寸
10 g
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
法律信息
Accuris is a trademark of Benchmark Scientific, Inc.
历史批次信息供参考:
分析证书(COA)