主要文件
Z531413
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 300 mm, joint: ST/NS 24/40
登录查看公司和协议定价
选择尺寸
1 EA
$310.00
$310.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$310.00
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
$310.00
请联系客服了解存货情况
接合处
joint: ST/NS24/40
存货情况
not available in EU
总高度
300 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)