主要文件
Z748250
Sigma-Aldrich
Load flask for Aldrich® Kugelrohr apparatus
capacity 250 mL, joint: ST/NS 24/40
别名:
distillation apparatus, distilling apparatus, fractional distillation apparatus, glass distillation apparatus, kugelrohr apparatus, kugelrohr distillation, short path distillation apparatus, vacuum distillation apparatus
登录查看公司和协议定价
选择尺寸
1 EA
$284.00
$284.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$284.00
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
$284.00
请联系客服了解存货情况
材料
glass
single-neck
spherical
接合處
joint: ST/NS24/40
無菌
non-sterile
特點
closure type Standard Taper Joint
包裝
case of 1
製造商/商標名
Sigma-Aldrich
容量
250 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
Heavy wall glass flask with solid support that attaches to Kugelrohr motor drive coupling. Fill load flask one-third full with distillable material.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)