主要文件
Z743171
Julabo® bath covers
flat cover to fit Pura 22, with 6 x 115 mm diam openings and set of rings
登录查看公司和协议定价
选择尺寸
1 EA
$536.00
$536.00
预计发货时间2025年7月25日详情
选择尺寸
变更视图
1 EA
$536.00
About This Item
UNSPSC代码:
41103706
NACRES:
NB.77
$536.00
预计发货时间2025年7月25日详情
包装
pack of 1 ea
制造商/商品名称
Julabo 9970574
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Flat bath cover for PURA 22 water bath, with 6 openings 115 mm diameter and corresponding ring sets. Place beakers or Erlenmeyer flasks directly on the stainless steel base plate.
法律信息
Julabo is a registered trademark of Julabo USA, Inc.
历史批次信息供参考:
分析证书(COA)