主要文件
Z743132
Julabo® cryo-compact circulator
CF30, capacity 3.5 L, temperature -30-150 °C, precision: ±0.03, AC/DC input 230 V AC, Schuko plug
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103706
NACRES:
NB.77
包装
pack of 1 ea
制造商/商品名称
Julabo 9400330
温度测量范围
-30-150 °C, precision: ±0.03
容量
3.5 L
正在寻找类似产品? 访问 产品对比指南
一般描述
The new CF series offers compact refrigerated/heating circulators. With small overall dimensions the units can even be placed on a very small surface or within a complex installation. All models offer a heating capacity of 2 kW and safety class 3 according to DIN12876-1. The maximum ambient temperature of +40°C as well as a ventilation air cooling allow installing the units very closely side by side with other units or directly under a fume hood. Furthermore all the models feature pump connections for external temperature applications and a bath opening for temperature control of small objects directly in the circulator bath. Wetted parts are made of high quality stainless steel or plastic. The instruments provide a reliable Microprocessor electronics with high temperature stability as well as warning and safety functions.
法律信息
Julabo is a registered trademark of Julabo USA, Inc.
历史批次信息供参考:
分析证书(COA)