主要文件
Z742608
Sigma-Aldrich
Photo KitAlysis 24-Green LED Array
登录查看公司和协议定价
About This Item
UNSPSC代码:
12352300
NACRES:
NA.22
一般描述
Green LED Array for Photo KitAlysis high-throughput screening platform. For use with Photo KitAlysis Starter Kit (Z742612). User guide is provided in the below hyperlink.
Photo KitAlysis Operating Instructions
Features:
  • Designed and tested by synthetic chemists.
  • Controller provides repeatable milliamp selection for photon intensity (sold seperately)
  • 527 nm wavelength
  • 0-30 mA variable LED output
  • Non-magnetic LED base
  • Chemically resistant LED cover
  • PTFE coated cabling
Photo Kitalysis Starter Kit required for operation (sold separately). Best when used with KitAlysis Benchtop Inertion Box (sold separately).
法律信息
KitAlysis is a trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)
访问文档库
商品
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
实验方案
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit
相关内容
The Photo KitAlysis Photoreactor Controller allows you maximum control of photocatalysis reactions.
The PhotoKitAlysis Reaction Kit was designed to provide the best possible chance of finding good reaction conditions for cross-coupling reactions: pre-weighed base or base solution (provided), 10–20 mmol coupling partner (user supplied), 10 mmol aryl halide substrate (user supplied), Catalysts (user supplied), 6 solvents (toluene, DMA. THJF, n-butanol, MeOH, MeCN) (provided).