主要文件
Z742529
Mini Nutating Rocker with flat mat, platform 8 x 6 in.
AC/DC input 115 V AC, US 3-pin plug
登录查看公司和协议定价
选择尺寸
1 EA
$819.00
$819.00
预计发货时间2025年5月30日详情
选择尺寸
变更视图
1 EA
$819.00
About This Item
UNSPSC代码:
41103801
$819.00
预计发货时间2025年5月30日详情
包装
pkg of 1
制造商/商品名称
Benchmark Scientific H3D1020
平台尺寸
8 in. × 6 in.
正在寻找类似产品? 访问 产品对比指南
一般描述
The Mini Nutating Rocker is pictured inside the MyTemp incubator; (supplied with both flat and dimpled mats) for Benchmark MyTemp Mini Digital Incubator H2200-H and H2200-HC
历史批次信息供参考:
分析证书(COA)
访问文档库
Questions
  1. What temperature range is this product rated for?
    1 answer
    1. The temperature range is +4C to +50C.