Merck
Z740687
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 2000 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
borosilicate glass flask
flat bottom flask
接合處
joint stopper9
包裝
pkg of 4 ea
容量
2000 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC