主要文件
Z691453
Scienceware® compact support stand
center and side mounts, base W × L 8 1/4 in. × 6 1/4 in., rod diam. × L 3/8 in. × 19 3/4 in.
登录查看公司和协议定价
选择尺寸
1 EA
$153.00
$153.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$153.00
About This Item
UNSPSC代码:
41122800
NACRES:
NB.35
$153.00
请联系客服了解存货情况
物料
polypropylene base
stainless steel rod
制造商/商品名称
Bel-Art H18303-0005
底座宽度 × 长度
8 1/4 in. × 6 1/4 in.
棒直径× 长度
3/8 in. × 19 3/4 in.
一般描述
This non-corrosive, molded support stand with stainless steel rod saves bench space. Lighter and less expensive than metal ring stands, it can be used to support a variety of types of plasticware and labware. The compact base has a smooth surface for easy cleaning and weighs 1.95 kg (4.3 lb) providing excellent stability. The stainless steel rod can be mounted at the center or at the end of the base. The base is made of corrosion-resistant polypropylene and is autoclavable.
法律信息
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)