主要文件
Z682071
Ace cold trap outer body only for condenser "C" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
登录查看公司和协议定价
选择尺寸
1 EA
$1,650.00
$1,650.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,650.00
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
$1,650.00
请联系客服了解存货情况
物料
plastic-coated glass
制造商/商品名称
Büchi equivalent, 40643
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
The cold trap outer body for the Ace condenser "C" assembly, is glass with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks. Features standard top and side flange that connects directly to the Büchi rotary evaporator.
法律信息
Büchi is a registered trademark of Büchi Labortechnik AG
历史批次信息供参考:
分析证书(COA)