主要文件
Z681679
PureSolv Micro vacuum pump/inert gas connection for multiple systems
pkg of 1 ea
登录查看公司和协议定价
选择尺寸
1 EA
$1,010.00
$1,010.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,010.00
About This Item
UNSPSC代码:
41100000
NACRES:
NB.85
$1,010.00
请联系客服了解存货情况
物料
anodized aluminum frame
stainless steel tubing
包装
pkg of 1 ea
制造商/商品名称
Innovative Technologies
一般描述
Two tees are required per Pure Solv Micro system when ganging vacuum and gas lines.
To connect 2 systems together you need qty 1 of this product
To connect 3 systems together you need qty 2 of this product
To connect 4 systems together you need qty 3 of this product
To connect 5 systems together you need qty 4 of this product
法律信息
Pure Solv is a trademark of Inert Corp.
历史批次信息供参考:
分析证书(COA)