主要文件
Z678759
FORTUNA® Polyfix® bottle top dispenser with PTFE coated piston
dispensing volume 10-50 mL
登录查看公司和协议定价
选择尺寸
1 EA
$403.00
$403.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$403.00
About This Item
UNSPSC代码:
41121500
NACRES:
NB.01
$403.00
请联系客服了解存货情况
制造商/商品名称
Fortuna Graf 101.071-48
dispensing volume measuring range
10-50 mL
体积
10-50 mL
配件
GL 40
GL 45
正在寻找类似产品? 访问 产品对比指南
一般描述
For dispensing of weak acids and alkaline solutions
Features:
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with PP-protection against breakage
  • Coated PTFE-piston
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • FIX adapter made of PP
  • Valve block made of PP, no metal springs in the valve - no corrosion
Supplied as FORTUNA Polyfix dispenser, 2 PP adapters GL40 + GL45 & 1 PTFE intake tube
法律信息
FORTUNA is a registered trademark of Poulten & Graf Ltd.
POLYFIX is a registered trademark of Poulten & Graf Ltd.
历史批次信息供参考:
分析证书(COA)