主要文件
Z678295
Finnpipette® F2 variable volume pipette
volume 1-10 μL , Micro-tip
登录查看公司和协议定价
选择尺寸
1 EA
$846.00
$846.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$846.00
About This Item
UNSPSC代码:
41121500
NACRES:
NB.01
$846.00
请联系客服了解存货情况
特点
color-coded pink
electronic: no
single-channel
包装
pack of 1 ea
制造商/商品名称
Finn 4642030
体积
1-10 μL , Micro-tip
正在寻找类似产品? 访问 产品对比指南
一般描述
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50μl and below
法律信息
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
历史批次信息供参考:
分析证书(COA)