主要文件
Z675547
Polypropylene carboy with spigot
size 20 L
别名:
laboratory dispenser
登录查看公司和协议定价
选择尺寸
1 EA
$367.00
$367.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$367.00
About This Item
分類程式碼代碼:
41121820
NACRES:
NB.35
$367.00
请联系客服了解存货情况
包裝
pack of 1 ea
製造商/商標名
Bel-Art F11846-0050
尺寸
20 L
正在寻找类似产品? 访问 产品对比指南
一般說明
Heavy-duty carboys have a molded-in, threaded bung that receives the spigot. All polypropylene construction is autoclavable 121°C (250°F) and chemical resistant. When autoclaving, do not install cap.
  • Single-handed spigot control for simple dispensing
  • 83 mm cap seals with a silicone gasket
  • Two large handles for transport or pouring
历史批次信息供参考:
分析证书(COA)