主要文件
Z563064
Work station anti-static ionizer for Plas-Labs glove box
AC/DC input 120 V AC
别名:
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
登录查看公司和协议定价
选择尺寸
1 EA
$1,750.00
$1,750.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,750.00
About This Item
UNSPSC代码:
41103406
NACRES:
NB.35
$1,750.00
请联系客服了解存货情况
物料
electrode (Thoriated tungsten)
制造商/商品名称
Plas-Labs 800-AS/SPI
存货情况
not available in EU
Overall H × W × depth
5.0 in. × 4 1/4 in. × 2 1/2 in.
重量
2 lb
正在寻找类似产品? 访问 产品对比指南
一般描述
The anti-static ionizer effectively eliminates all static charges within 36 inches of the unit and is useful in any Plas-Labs glove box or isolation chamber.
Operation is simple; place the unit on a flat surface and turn the power on.
Unit features include: ion pulse rate adjustment from 1 to 5 pulses/second, ion balance adjustment and a compact footprint. The unit is virtually maintenance free; the emitter points should be cleaned when a visual accumulation of dust or powder occurs.
法律信息
历史批次信息供参考:
分析证书(COA)