主要文件
Z549339
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 100 mL
登录查看公司和协议定价
选择尺寸
1 EA
$284.00
$284.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$284.00
About This Item
UNSPSC代码:
41121815
NACRES:
NB.35
$284.00
请联系客服了解存货情况
接合处
female joint: ST/NS29/32
male joint: ST/NS24/40
容量
100 mL
孔径
70-100 μm porosity
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)