主要文件
Z530697
Sigma-Aldrich
Aldrich® vacuum manifold with joints
positions, 3, joint: ST/NS 24/40
登录查看公司和协议定价
选择尺寸
1 EA
$1,410.00
$1,410.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,410.00
About This Item
UNSPSC代码:
40161501
NACRES:
NB.79
$1,410.00
请联系客服了解存货情况
接合处
joint: ST/NS24/40
特点
positions 3
长度
300 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
This practical design provides clearance to accommodate 250mL flasks with a snap of a Keck® clip. Manifolds are available with either <ST/>14/20, <ST/>24/40, or <ST/>29/32 joints. Glass stopcocks have a 4mm bore.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Keck is a registered trademark of Adcuram Lab Technologies GmbH
历史批次信息供参考:
分析证书(COA)