主要文件
Z503673
Sigma-Aldrich
Scienceware® rotary pipette stand
holds up to 94 pipettes and swivels for easy access
登录查看公司和协议定价
选择尺寸
1 EA
$154.00
$154.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$154.00
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
$154.00
请联系客服了解存货情况
包裝
pack of 1 ea
製造商/商標名
Bel-Art F18957-0000
一般說明
PP and PVC rack holds up to 94 pipettes and swivels for easy access. Base has raised lip to retain draining liquid. Unit stands 48 cm high when assembled, and the rotating disk diameter is 18 cm.
法律資訊
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)