主要文件
Z422916
Sigma-Aldrich
Aldrich® pressure filter/drying funnel assemblies with removable fritted disc
funnel capacity 500 mL
登录查看公司和协议定价
选择尺寸
1 EA
$1,360.00
$1,360.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,360.00
About This Item
UNSPSC代码:
41121809
NACRES:
NB.24
$1,360.00
请联系客服了解存货情况
物料
glass
制造商/商品名称
Aldrich
漏斗容量
500 mL
孔径
coarse
容量
500 mL capacity
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Pressure or vacuum can be applied alternately or simultaneously for fast filtration or drying. Unique design allows the fritted disc to be removed and replaced. The bottom support is all PTFE and has a vent. A port is supplied on the top cap for an optional adjustable pressure relief valve. Large top and bottom openings are 85 mm I.D. allowing easy access to funnel contents. Complete assembly includes top cap, funnel, coarse frit, 90 mm base assembly, and 2 L bottle with GL 45 thread.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)
访问文档库
其他客户在看
Slide 1 of 1
1 of 1
Questions
  1. Hi, This set-up seems exactIy what I'm looking for but can't seem to find the pressure rating. What pressure can be applied to this? Best regards, Carl
    1 answer
    1. Max pressure is 15psi.