主要文件
Z418145
Sigma-Aldrich
Aldrich® System 45 powder addition funnel
capacity 125 mL
别名:
funnel powder, powder funnel
登录查看公司和协议定价
选择尺寸
1 EA
$742.00
$742.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$742.00
About This Item
UNSPSC代码:
41121809
NACRES:
NB.43
$742.00
请联系客服了解存货情况
螺旋钻尺寸
large
接合处
joint ST/NS24/40
高度
~250 mm
容量
125 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
These funnels are designed to deliver free-flowing granular material to reactions by turning the auger feed. Delivery can be continuous or a single charge. Funnels are designed with large top openings with GL 45 threaded closures. PTFE auger can be removed easily for cleaning and is designed for vacuum applications. Only the 500 mL size has a clean out side-arm port. Supplied complete with auger and top cap.
其他说明
When assembling the auger into the remaining equipment, the auger unit must be fully advanced past the glass threads. Use water to moisten the o-ring when necessary. Use proper personal protective equipment (PPE) when handling glassware.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
替代部件
历史批次信息供参考:
分析证书(COA)