主要文件
Z285463
Sigma-Aldrich
PowrTrol heat controllers
AC/DC input 120 V AC, 10 A
登录查看公司和协议定价
About This Item
UNSPSC代码:
41112303
NACRES:
NB.76
价格与库存信息目前不能提供
制造商/商品名称
Glas-Col 104APL120
存货情况
not available in EU
功耗
1,200 W
正在寻找类似产品? 访问 产品对比指南
一般描述
  • PowrTrol heat controller is a solid-state, proportional-voltage power control that provides precise manual control of mantles and tapes.
  • The output is adjustable from 3 to 100% of rated voltage.
  • The unit can be mounted to a lattice or used on bench top.
PowrTrol heat controllers includes:
Lighted On/Off switch, 10-amp output, Non-Linear control (scale is for reference only), Circuit protection, Low profile housing.
A solid-state, proportional-voltage power control that provides precise manual control of mantles and tapes. The output is adjustable from 3 to 100% of rated voltage. Can be mounted to a lattice or used on bench top.
历史批次信息供参考:
分析证书(COA)
访问文档库
其他客户在看
Slide 1 of 1