主要文件
Z278351
ACE Micro/Mini-Lab® Hickman-Hinkle stillhead
joint: ST/NS 14/10 top and bottom, head size 10 mL
登录查看公司和协议定价
选择尺寸
1 EA
$158.00
$158.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$158.00
About This Item
UNSPSC代码:
41100000
NACRES:
NB.43
$158.00
请联系客服了解存货情况
接合处
joint: ST/NS14/10 top and bottom
制造商/商品名称
Ace Glass 959962
头部尺寸
10 mL
相关类别
一般描述
For conducting simple and fractional distillations. Has threaded side port with septum and cap for syringe access to well contents and an elongated section below well to accommodate the PTFE spinning band. 10 mL still has 14/10 threaded outer joint at top and 14/10 inner joint at bottom. Top outer joint supplied with cap, septum, and O-ring.
法律信息
Micro/Mini-Lab is a registered trademark of Ace Glass, Inc.
历史批次信息供参考:
分析证书(COA)