主要文件
Z268755
Scienceware® Safe-Lab PTFE stirrer bearing
joint: ST/NS 29/42
登录查看公司和协议定价
选择尺寸
1 EA
$440.00
$440.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$440.00
About This Item
UNSPSC代码:
41100000
NACRES:
NB.72
$440.00
请联系客服了解存货情况
物料
Viton® O-ring
接合处
joint: ST/NS29/42
包装
pack of 1 ea
制造商/商品名称
Bel-Art F20320-1662
正在寻找类似产品? 访问 产品对比指南
一般描述
Stopper-Loc® nut allows stirrer bearing to be locked to outer ground-glass joint. Complete with Stopper-Loc nut and two PTFE inserts. For 10mm diam. polished glass or PTFE stirring shafts.
法律信息
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Stopper-Loc is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
历史批次信息供参考:
分析证书(COA)