主要文件
Z261793
Sigma-Aldrich
Manual gas control valve
Monel®, CGA Inlet size 330, barb for 1/4in. i,d. hose
登录查看公司和协议定价
About This Item
UNSPSC代码:
41112404
NACRES:
NB.51
价格与库存信息目前不能提供
物料
Monel®
入口压力测量范围
≤3,000 psig
CGA入口尺寸
330
正在寻找类似产品? 访问 产品对比指南
一般描述
Provides an economical means of controlling gas flow directly from the cylinder. Manufactured with SS or Monel® bodies to accommodate corrosive gases that cannot be run through a regulator. Valve stem packing is PTFE. Hose barb outlet.
警告
Control valves do not regulate pressure, they only control flow.
法律信息
Monel is a registered trademark of Huntington Alloys Corp.
历史批次信息供参考:
分析证书(COA)