主要文件
Z222364
Metal standard taper clip
joint size, 14
登录查看公司和协议定价
选择尺寸
6 EA
$141.00
$141.00
请联系客服了解存货情况
选择尺寸
变更视图
6 EA
$141.00
About This Item
分類程式碼代碼:
41122413
NACRES:
NB.35
$141.00
请联系客服了解存货情况
一般說明
Nickel-plated spring-steel joint clips offer increased strength and resistance to heat and cracking over Delrin® plastic type. Securely holds conical joints under pressure at temperatures up to 392 °F (200 °C). Will not scratch glassware.
法律資訊
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
历史批次信息供参考:
分析证书(COA)