主要文件
Z217530
Flexible stirrer shaft coupling
for glass shaft O.D. 10 mm, for motor shaft O.D. 3/8 in.
登录查看公司和协议定价
选择尺寸
1 EA
$131.00
$131.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$131.00
About This Item
UNSPSC代码:
41100000
NACRES:
NB.72
$131.00
请联系客服了解存货情况
正在寻找类似产品? 访问 产品对比指南
一般描述
Flexible coupling allows angular and parallel misalignment of glass stirring shaft without breakage. Neoprene rubber sleeve body absorbs shock and vibration and has great torsional stiffness for quiet, smooth running. PTFE sleeve slides over end of glass shaft. SS set screw secures other end of coupling to motor shaft.
历史批次信息供参考:
分析证书(COA)