主要文件
Z202789
Sigma-Aldrich
Aldrich® adjustable rapid-purge valve
登录查看公司和协议定价
选择尺寸
1 EA
$1,350.00
$1,350.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,350.00
About This Item
UNSPSC代码:
40141600
NACRES:
NB.79
$1,350.00
请联系客服了解存货情况
相关类别
一般描述
Designed for mounting anywhere in a vacuum system. Use for purging evacuated apparatus with inert gas. Two-piece PTFE plug with Viton® O-ring and SS spring is adjustable for pressures of 5 to 35 psi.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Viton is a registered trademark of E. I. du Pont de Nemours and Company
历史批次信息供参考:
分析证书(COA)
访问文档库
其他客户在看
Slide 1 of 1