主要文件
Z190268
Arrow electric lab stirrer
speed 0-850 rpm, gear, output0.10 hp
别名:
lab mixer, lab mixers, mixer stirrer
登录查看公司和协议定价
选择尺寸
1 EA
$1,910.00
$1,910.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,910.00
About This Item
UNSPSC代码:
41103801
NACRES:
NB.72
$1,910.00
请联系客服了解存货情况
AC/DC输入
115 V
115 V
包装
pack of 1 ea
制造商/商品名称
Arrow Engineering 850
存货情况
not available in EU
参数
0-850 rpm speed
输出
0.10 hp
正在寻找类似产品? 访问 产品对比指南
一般描述
Totally enclosed, non-ventilated motor seals out corrosive fumes and dust. With on/off switch, circuit-breaker, and variable speed control. Includes a 9/16 × 10 in. support rod and 3/8 diam. × 12 in. L SS shaft and propeller.
质量
Note: Arrow lab stirrers conform to CSA22.2 but are not certified
历史批次信息供参考:
分析证书(COA)