主要文件
Z180947
Ace auto-zero McLeod vacuum gauges
pressure 0-10 mmHg
登录查看公司和协议定价
选择尺寸
1 EA
$2,420.00
$2,420.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$2,420.00
About This Item
分類程式碼代碼:
41112406
NACRES:
NB.79
$2,420.00
请联系客服了解存货情况
製造商/商標名
Ace Glass 872551
壓力測量範圍
0-10 mmHg
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
A tilting gauge with a constant zero overflow which eliminates the capillary-depression error from an expanding meniscus. Once the zero line of the scale is aligned with the tip of the closed capillary, it is only necessary to bring the gauge to an upright position and read the scale behind the closed leg.
Gauges are fabricated from calibrated ACE Trubore tubing, to assure uniform and reproducible reading between the gauges. Mercury trap is incorporated in female joint, bulbs are enlarged. Joint is standard taper 20/65.
The pressure range is: 0-1.0 mmHg and the lowest reading is: 0.001 mmHg.
Mercury not included (approximately 20 mL of triple distilled mercury is needed to ffill this vacuum gauge to working volume).
历史批次信息供参考:
分析证书(COA)