主要文件
Z173495
Sigma-Aldrich
Addition funnel with Rotaflo® stopcock, and glass stopcock on side-arm
capacity 100 mL, male joint: ST/NS 14/20, female joint: ST/NS 14/20
登录查看公司和协议定价
选择尺寸
1 EA
$542.00
$542.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$542.00
About This Item
UNSPSC代码:
41121815
NACRES:
NB.43
$542.00
请联系客服了解存货情况
标签
W
接合处
female joint: ST/NS14/20
male joint: ST/NS14/20
容量
100 mL
相关类别
法律信息
Rotaflo is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)
访问文档库
商品
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.