Merck
Z152412
Stainless steel 304 syringe needle
gauge 18, L 36 in.
登录查看公司和协议定价
选择尺寸
1 EA
$141.00
$141.00
预计发货时间2025年4月21日详情
选择尺寸
变更视图
1 EA
$141.00
About This Item
UNSPSC代码:
41122002
NACRES:
NB.03
$141.00
预计发货时间2025年4月21日详情
描述
Standard Hub with 304 Stainless Steel Tubing
包装
pack of 1 ea
制造商/商品名称
Cadence Science
长度
36 in.
计量器
18
正在寻找类似产品? 访问 产品对比指南
一般描述
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)