主要文件
Z147923
Sigma-Aldrich
Glass sample holder
Boat, size 2 1/2 in. × 10 in.
登录查看公司和协议定价
选择尺寸
1 EA
$197.00
$197.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$197.00
About This Item
分類程式碼代碼:
41100000
NACRES:
NB.43
$197.00
请联系客服了解存货情况
材料
borosilicate glass
參數
230-490 °C temp. limit
尺寸
2 1/2 in. × 10 in.
正在寻找类似产品? 访问 产品对比指南
一般說明
These sample holders come in both boat and tube designs, made of borosilicate glass. The glass vessel is ideal for transporting and drying chemical samples in the Aldrich Abderhalden apparatus, in desiccators, or in any drying oven. It has a molded end piece to grip and easier handling of the boat.
历史批次信息供参考:
分析证书(COA)