主要文件
WHAMUPMCPBC8
Whatman® Mini-UniPrep® syringeless filters accessories
Multicompressor unit
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104922
NACRES:
NB.24
产品WHAMUPMCPBC8监管中,请联系销售询价或购买 联系客户支持
无菌性
non-sterile
特点
Multicompressor unit
制造商/商品名称
Whatman MUPMCPBC8
正在寻找类似产品? 访问 产品对比指南
一般描述
Contains a Multicompressor and a single 8 position blue tray to be used with plastic MUP
特点和优势
  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva′s Mini-UniPrepstandard plastic filter vial
  • Mini-UniPrepvials can be loaded directly into the autosampler
法律信息
Uniprep is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva
历史批次信息供参考:
分析证书(COA)