主要文件
TEM-76042-40
Sigma-Aldrich
Micronporous Silicon Nitride TEM Window Grids for Cryo-EM Suspension
10nm Thick, pack of 10
登录查看公司和协议定价
About This Item
UNSPSC代码:
41111761
NACRES:
MA.24
产品TEM-76042-40监管中,请联系销售询价或购买 联系技术服务部门
包装
pack of 10
正在寻找类似产品? 访问 产品对比指南
一般描述
Silicon nitrode microporous TEM window with single square of 500x500 micron size, and 20 nm thick SiN. Plasma cleanable, less than 0.5nm variation in film thickness, tolerate temperatures above 1000 ˚C, provides flat, insulating and hydrophobic surfaces
历史批次信息供参考:
分析证书(COA)