主要文件
SYNF804260
Synthware Buchner funnel with perforated plate
60 mL, plate diam. 42 mm
别名:
Synthware F804260, filtering funnel
登录查看公司和协议定价
选择尺寸
1 EA
$178.00
$178.00
预计发货时间2025年5月07日详情
选择尺寸
变更视图
1 EA
$178.00
About This Item
UNSPSC代码:
41121809
NACRES:
NB.51
$178.00
预计发货时间2025年5月07日详情
包装
pkg of 1 ea
制造商/商品名称
Synthware F804260
容量
60 mL
60 mL
板直径
42 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
With sealed-in perforated glass disc to support filter media. Pressure may be used but should not exceed 15 psi. This design solve the contamination problem with fritted disc and also the blindness problem with ceramic Buchner funnel.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)