主要文件
SYNF603030M
Synthware Buchner funnel
30 mL, porosity: medium
别名:
Synthware F603030M, filtering funnel
登录查看公司和协议定价
选择尺寸
1 EA
$67.10
$67.10
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$67.10
About This Item
UNSPSC代码:
41121809
NACRES:
NB.51
$67.10
请联系客服了解存货情况
包装
pkg of 1 ea
制造商/商品名称
Synthware F603030M
容量
30 mL
30 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)