主要文件
SYNF49081L
Synthware storage flask with high vacuum valve and side arm
1000 mL, valve size 0-8 mm
别名:
Synthware F49081L, round bottom flask
登录查看公司和协议定价
选择尺寸
1 EA
$215.00
$215.00
预计发货时间2025年6月06日详情
选择尺寸
变更视图
1 EA
$215.00
About This Item
UNSPSC代码:
41121804
NACRES:
NB.51
$215.00
预计发货时间2025年6月06日详情
物料
borosilicate glass flask
round bottom
single-neck (high vacuum valve)
包装
pkg of 1 ea
制造商/商品名称
Synthware F49081L
容量
1,000 mL
1000 mL
1000 mL
阀门尺寸
0-8 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper( “umbrella”) to protect Viton® O-ring on shaft..
法律信息
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
历史批次信息供参考:
分析证书(COA)