主要文件
SYNA301410
Synthware gas inlet/outlet adapter
joint: ST/NS 14/10
别名:
Synthware A301410, inert gas adapter, vacuum adapter
登录查看公司和协议定价
选择尺寸
1 EA
$30.60
$30.60
预计发货时间2025年6月02日详情
选择尺寸
变更视图
1 EA
$30.60
About This Item
UNSPSC代码:
41121800
NACRES:
NB.51
$30.60
预计发货时间2025年6月02日详情
接合处
joint: ST/NS14/10
包装
pack of 1 ea
制造商/商品名称
A301410
相关类别
一般描述
Synthware gas inlet/outlet adapter is used to facilitate the introduction and removal of gases in various system. The joint is 14/10 and the hose connection is 8 mm O.D.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)