跳转至内容
Merck
所有图片(1)

主要文件

PH003193

Sigma-Aldrich

2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propanamide

AldrichCPR

登录查看公司和协议定价


About This Item

线性分子式:
C11H10N2O3
CAS号:

SMILES字符串

O=C(C1=C2C=CC=C1)N(C(C)C(N)=O)C2=O

InChI

1S/C11H10N2O3/c1-6(9(12)14)13-10(15)7-4-2-3-5-8(7)11(13)16/h2-6H,1H3,(H2,12,14)

其他说明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

象形图

Exclamation mark

警示用语:

Warning

危险声明

危险分类

Acute Tox. 4 Oral - Eye Irrit. 2

储存分类代码

11 - Combustible Solids

WGK

WGK 3

闪点(°F)

Not applicable

闪点(°C)

Not applicable


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

Questions

Reviews

No rating value

Active Filters

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门