主要文件
CLS69474L
Pyrex® resin reaction kettles
capacity 4,000 mL
别名:
pyrex flask, pyrex with lid, pyrex with lids, Corning® 6947
登录查看公司和协议定价
选择尺寸
1 EA
$527.00
$527.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$527.00
About This Item
UNSPSC代码:
41121804
$527.00
请联系客服了解存货情况
包装
case of 1 ea
制造商/商品名称
Corning 6947-4L
存货情况
not available in EU
容量
4,000 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Designed for viscous reaction materials. 500 and 1,000 mL kettles have interchangeable covers with four openings (24/40 joints). 2,000, 3,000 and 4,000 mL kettles have three openings with 24/40 joints and one with a 34/45 joint. Flanges of covers and bottoms are finely ground for a tight seal. Order clamp separately.
法律信息
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)