主要文件
CLS6795611
Corning® 11in x 11in stirrer, model PC-611
AC/DC input 120 V AC, plate W × L 11 in. × 11 in.
别名:
magnetic stirrer, magnetic stirrers, stirrers
登录查看公司和协议定价
选择尺寸
1 EA
$832.00
$832.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$832.00
About This Item
UNSPSC代码:
41103803
NACRES:
NB.73
$832.00
请联系客服了解存货情况
制造商/商品名称
Corning 6795-611
存货情况
not available in EU
stirring range measuring range
100-1500 rpm
板宽度 × 长度
11 in. × 11 in.
一般描述
These Corning PC-611 stirrers have an 11 x 11 inch plastic (ABS) top and can stir large volumes of liquid with ease. These stirrers are designed for 5 gallon vessels (19L), but can stir up to 10 gallons and can handle many applications that previously required an overhead stirrer. This powerful stirring is achieved with a very compact design that conserves bench space. The stirrer uses a special rare earth magnet that is constructed out of for exceptional coupling strength. The magnet′s strength also centers the stir bar quickly inside the vessel. They operate on 120V/60Hz and are UL and CUL approved to meet needs in North America and other locations with similar requirements.
Heavy duty motor and rugged plastic (ABS) top will provide years of reliable service. -More compact and lower cost than competitive units. -UL/CUL approved. -1 year warranty.
法律信息
Corning is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)