Merck
CLS4965100
Pyrex® round-bottom three neck angle type distilling flask
capacity 100 mL
别名:
bottom flask, flask round, flask round bottom
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41104812
材料
borosilicate glass
round bottom flask
three-neck (Vertical)
接合處
center joint ST/NS24/40
side joint ST/NS19/38
無菌
non-sterile
特點
closure type Standard Taper joint
包裝
pack of 1
case of 4
製造商/商標名
Corning 4965-100
容量
100 mL
正在寻找类似产品? 访问 产品对比指南
法律資訊
PYREX is a registered trademark of Corning, Inc.