主要文件
CAD6219
1-way tubing connector
MLL to Tuohy Borst (1-5 French) (plated brass)
登录查看公司和协议定价
选择尺寸
10 EA
$1,030.00
$1,030.00
请联系客服了解存货情况
选择尺寸
变更视图
10 EA
$1,030.00
About This Item
UNSPSC代码:
40183110
NACRES:
NB.51
$1,030.00
请联系客服了解存货情况
包装
pack of 10 ea
制造商/商品名称
Cadence Science 6219
正在寻找类似产品? 访问 产品对比指南
一般描述
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
法律信息
Micro-Mate is a registered trademark of Cadence Science
历史批次信息供参考:
分析证书(COA)